Skip to Content
Merck
All Photos(2)

Key Documents

778087

Sigma-Aldrich

Molybdenum(IV) selenide

-325 mesh, 99.9% trace metals basis

Synonym(s):

Molybdenum diselenide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MoSe2
CAS Number:
Molecular Weight:
253.86
EC Number:
MDL number:
UNSPSC Code:
12352300
NACRES:
NA.23

Assay

99.9% trace metals basis

form

powder

particle size

-325 mesh

mp

>400 °C

application(s)

battery manufacturing

SMILES string

[Mo].[Se].[Se]

InChI

1S/Mo.2Se

InChI key

MHWZQNGIEIYAQJ-UHFFFAOYSA-N

Application

Molybdenum chalcogenide thin film transitors for solar cells are a rapidly expanding area of research and molybdenum selenide compounds play an important role in this research. The layered nature of MoSe2 gives rise to several interesting applications such as field effect transistors ; piezoelectrics and substrates for self assembly

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Duerloo; K.N.;
The Journal of Physical Chemistry Letters, 3(19), 2871-2876 (2012)
Larentis; S.;
Applied Physics Letters, 101(22), 223104/1-223104/4 (2012)
Abou-Ras; D.;
Thin Solid Films, 480-481, 433-438 (2005)
Kohara; N.;
Solar Energy Materials and Solar Cells, 67(1-4), 209-215 (2001)
Cincotti; S.;
Applied Physics Letters, 62(26), 3531-3533 (1993)

Articles

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service