Skip to Content
Merck
All Photos(1)

Documents

777064

Sigma-Aldrich

1-Octen-3-one

96%

Synonym(s):

Amyl vinyl ketone, Pentyl vinyl ketone, Vinyl amyl ketone, n-Amyl vinyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O
CAS Number:
Molecular Weight:
126.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.435

density

0.833 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCCCCC(=O)C=C

InChI

1S/C8H14O/c1-3-5-6-7-8(9)4-2/h4H,2-3,5-7H2,1H3

InChI key

KLTVSWGXIAYTHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

164.8 °F - closed cup

Flash Point(C)

73.8 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Franitza et al.
Journal of agricultural and food chemistry, 64(47), 9041-9053 (2016-10-30)
The production of rum consists of fermentation, distillation, and aging. To check the influence of each step on the final rum aroma, molasses, mash, distillate, and the final rum were analyzed using the sensomics concept. The changes in key aroma
James Ridsdill-Smith et al.
Archives of insect biochemistry and physiology, 51(4), 170-181 (2002-11-15)
Halotydeus destructor feeding on subterranean clover cotyledons can cause severe damage. The mites live on the soil surface and move up onto plants to feed. Foraging behaviour consists of palpating, probing, and feeding with frequent transitions between them. Sustained feeding
Philippe Darriet et al.
Journal of agricultural and food chemistry, 50(11), 3277-3282 (2002-05-16)
Powdery mildew due to the fungus Uncinula necator is an important disease for the vineyard. The development of the fungus at the surface of the berries leads to the occurrence of a very characteristic and sometimes intense mushroom-type odor cited
Yong Cheng et al.
Journal of agricultural and food chemistry, 56(6), 2160-2169 (2008-02-27)
The temporal change in the headspace composition of an aroma model mimicking Longjing green tea aroma was studied in the presence of nonvolatile Longjing green tea constituents. Upon storage at 50 degrees C, the formation of 2-butyl-2-octenal was found, which
Meryl B Lubran et al.
Journal of agricultural and food chemistry, 53(21), 8325-8327 (2005-10-13)
Taste threshold tests of ferrous sulfate (FeSO4) solutions have been confounded by the presence of putative odorants. To detect the presence of odorants released from these solutions solid-phase microextraction (SPME) was used to collect volatiles in the headspace above FeSO4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service