Skip to Content
Merck
All Photos(1)

Key Documents

776262

Sigma-Aldrich

3,6-Bis(5-bromo-2-thienyl)-2,5-bis(2-hexyldecyl)-2,5-dihydro-pyrrolo[3,4-c]pyrrole-1,4-dione

98%

Synonym(s):

Pyrrolo[3,4-c]pyrrole-1,4-dione,3,6-bis(5-bromo-2-thienyl)-2-5-bis(2-hexyldecyl)-2,5-dihydro-

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H70Br2N2O2S2
CAS Number:
Molecular Weight:
907.00
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

solid

mp

110-115 °C

SMILES string

O=C1N(CC(CCCCCC)CCCCCCCC)C(C2=CC=C(Br)S2)=C3C1=C(C4=CC=C(Br)S4)N(CC(CCCCCC)CCCCCCCC)C3=O

InChI

1S/C46H70Br2N2O2S2/c1-5-9-13-17-19-23-27-35(25-21-15-11-7-3)33-49-43(37-29-31-39(47)53-37)41-42(45(49)51)44(38-30-32-40(48)54-38)50(46(41)52)34-36(26-22-16-12-8-4)28-24-20-18-14-10-6-2/h29-32,35-36H,5-28,33-34H2,1-4H3

InChI key

MNALLGNMYJQSHP-UHFFFAOYSA-N

General description

3,6-Bis(5-bromo-2-thienyl)-2,5-bis(2-hexyldecyl)-2,5-dihydro-pyrrolo[3,4-c]pyrrole-1,4-dione (DDP-Br2) belongs to the series of diketopyrrolopyrrole (DPP) based materials which show hole or ambipolar transport behavior with mobilities of charge carriers in the range of 0.1-1 cm2V-1 s-1. It has an electron deficient core that forms a low LUMO energy level which facilitates better air-stability for n-type organic semiconductors.

Application

DDP-Br2 can be used in the fabrication of a variety of opto-electronic devices such as organic field effect transistors (OFETs), perovskite based solar cells, non-fullerene organic solar cells.
Novel acceptors utilized in low band gap polymer cells and showed optical bandgaps ranging from 1.81 to 1.94 eV and intense absorption bands that cover a wide range from 300 to 700 nm

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Materials Chemistry, 21, 1600-1606 (2011)
A Novel BODIPY-Based Low-Band-Gap Small-Molecule Acceptor for Efficient Non-fullerene Polymer Solar Cells
Liu W, et al.
Chin. J. Chem., 35(12), 1813-1823 (2017)
Hole-transporting diketopyrrolopyrrole-thiophene polymers and their additive-free application for a perovskite-type solar cell with an efficiency of 16.3%
Maruo H, et al.
Polymer Journal, 51(1), 91-91 (2019)
Optimization of broad-response and high-detectivity polymer photodetectors by bandgap engineering of weak donor-strong acceptor polymers
Qi J, et al.
Macromolecules, 48(12), 3941-3948 (2015)
Journal of Materials Chemistry, 22, 2120-2128 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service