Skip to Content
Merck
All Photos(1)

Key Documents

77540

Sigma-Aldrich

o-Phenetidine

≥97.0% (GC)

Synonym(s):

2-Ethoxyaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H4NH2
CAS Number:
Molecular Weight:
137.18
Beilstein:
637069
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 67 °C)

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.555 (lit.)
n20/D 1.555

bp

231-233 °C (lit.)

density

1.051 g/mL at 25 °C (lit.)

SMILES string

CCOc1ccccc1N

InChI

1S/C8H11NO/c1-2-10-8-6-4-3-5-7(8)9/h3-6H,2,9H2,1H3

InChI key

ULHFFAFDSSHFDA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fan Xiao et al.
Laboratory investigation; a journal of technical methods and pathology, 97(3), 289-301 (2016-12-20)
Nonalcoholic fatty liver disease is considered a disorder of the endoplasmic reticulum (ER) and mitochondria. Recent studies have shown that the ER and mitochondrial membranes overlap by 15-20%, a region referred to as the 'mitochondria-associated ER membrane' (MAM). Some proteins
Karina Ferreira Noronha Cruz et al.
Journal of biomedical materials research. Part A, 105(2), 601-607 (2016-10-12)
Electroactive nanofibers based on thermoplastic polyurethane (TPU) and poly(alkoxy anilines) produced by electrospinning has been explored for biomaterials applications. The thermoplastic polyurethane is a biocompatible polymer with good mechanical properties. The production of TPU nanofibers requires the application of high
Christophoros Mannaris et al.
Advanced healthcare materials, 7(12), e1800184-e1800184 (2018-04-27)
The efficient penetration of drugs into tumors is a major challenge that remains unmet. Reported herein is a strategy to promote extravasation and enhanced penetration using inertial cavitation initiated by focused ultrasound and cone-shaped gold nanoparticles that entrap gas nanobubbles.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service