757454
(S)-3-Aminobutyric acid
97%
Synonym(s):
(S)-β-Aminobutyric acid, S-3-Aminobutanoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H9NO2
CAS Number:
Molecular Weight:
103.12
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
solid
mp
229-231 °C
functional group
amine
carboxylic acid
storage temp.
2-8°C
SMILES string
N[C@H](CC(O)=O)C
InChI
1S/C4H9NO2/c1-3(5)2-4(6)7/h3H,2,5H2,1H3,(H,6,7)/t3-/m0/s1
InChI key
OQEBBZSWEGYTPG-VKHMYHEASA-N
Related Categories
General description
(S)-3-Aminobutyric acid is a nonproteogenic amino acid.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yan-Lei Du et al.
Journal of experimental botany, 63(13), 4849-4860 (2012-08-04)
A pot experiment was conducted to investigate the effect of the non-protein amino acid, β-aminobutyric acid (BABA), on the homeostasis between reactive oxygen species (ROS) and antioxidant defence during progressive soil drying, and its relationship with the accumulation of abscisic
Comparative proteomic analysis of ?-aminobutyric acid-mediated alleviation of salt stress in barley.
Mostek A, et al.
Plant Physiology and Biochemistry, 99, 150-161 (2016)
Quantification of aminobutyric acids and their clinical applications as biomarkers for osteoporosis.
Zhiying Wang et al.
Communications biology, 3(1), 39-39 (2020-01-24)
Osteoporosis is a highly prevalent chronic aging-related disease that frequently is only detected after fracture. We hypothesized that aminobutyric acids could serve as biomarkers for osteoporosis. We developed a quick, accurate, and sensitive screening method for aminobutyric acid isomers and
Yajun Li et al.
Physiologia plantarum, 136(1), 19-29 (2009-06-11)
DL-beta-amino-butyric acid (BABA) has been found to protect potato plants against late blight caused by Phytophthora infestans. This paper compares gene expression profiles of a potato clone harboring horizontal resistance to P. infestans in response to BABA and the pathogen
Lennart Eschen-Lippold et al.
Molecular plant-microbe interactions : MPMI, 23(5), 585-592 (2010-04-07)
Inducing systemic resistance responses in crop plants is a promising alternative way of disease management. To understand the underlying signaling events leading to induced resistance, functional analyses of plants defective in defined signaling pathway steps are required. We used potato
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service