Skip to Content
Merck
All Photos(3)

Key Documents

755710

Sigma-Aldrich

Carbon nanotube, single-walled

>70% (TGA), avg. no. of layers, 1

Synonym(s):

NC-1100, SWCNT, SWNT, Single wall carbon nanotube

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

>70% (TGA)

form

powder

feature

avg. no. of layers 1

manufacturer/tradename

Nanocyl, Inc.

size L

× 3 μm , TEM

average diameter

2 nm

mp

3652-3697 °C (lit.)

density

1.7-1.9 g/cm3 at 25 °C (lit.)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

These single walled carbon nanotubes (SWNTs) were produced via the Catalytic Carbon Vapor Deposition (CCVD) process. Potential applications of SWNTs are in field effect transistors (FETs), solar cells, photodetectors, high performance computation, renewable energy, optoelectronics etc.

Application

Potential applications of SWNTs are in field effect transistors (FETs), solar cells, photodetectors, high performance computation, renewable energy, optoelectronics etc.Other applications include electronics,sensors,composites,energy storage, study of life science systems.

Preparation Note

Catalytic Carbon Vapor Deposition (CCVD) Method

Legal Information

Product of Nanocyl, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dominick J Bindl et al.
ACS nano, 4(10), 5657-5664 (2010-10-07)
Semiconducting single-walled carbon nanotubes (s-SWCNTs) have strong near-infrared and visible absorptivity and exceptional charge transport characteristics, rendering them highly attractive semiconductor absorbers for photovoltaic and photodetector technologies. However, these applications are limited by a poor understanding of how photogenerated charges
Wai Li Ling et al.
ACS nano, 5(2), 730-737 (2011-01-11)
The classical pathway of complement is an essential component of the human innate immune system involved in the defense against pathogens as well as in the clearance of altered self-components. Activation of this pathway is triggered by C1, a multimolecular
Amin Salehi-Khojin et al.
ACS nano, 5(1), 153-158 (2010-12-29)
There has been recent controversy whether the response seen in carbon nanotube (CNT) chemiresistors is associated with a change in the resistance of the individual nanotubes or changes in the resistance of the junctions. In this study, we carry out
Mohammad-Bagher Ebrahim-Habibi et al.
Scientific reports, 9(1), 1273-1273 (2019-02-06)
Investigation of non-covalent interaction of hydrophobic surfaces with the protein G (PrG) is necessary due to their frequent utilization in immunosensors and ELISA. It has been confirmed that surfaces, including carbonous-nanostructures (CNS) could orient proteins for a better activation. Herein

Articles

Find unique properties & applications of single (SWNTs) , double (DWNTs) & multi walled carbon nanotubes (MWCNTs).

Find unique properties & applications of single (SWNTs) , double (DWNTs) & multi walled carbon nanotubes (MWCNTs).

Find unique properties & applications of single (SWNTs) , double (DWNTs) & multi walled carbon nanotubes (MWCNTs).

Find unique properties & applications of single (SWNTs) , double (DWNTs) & multi walled carbon nanotubes (MWCNTs).

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service