Skip to Content
Merck
All Photos(1)

Documents

750905

Sigma-Aldrich

Metalaxyl-(phenyl-13C6)

99 atom % 13C, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C9H21NO4
CAS Number:
Molecular Weight:
285.29
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.24

isotopic purity

99 atom % 13C

Quality Level

Assay

98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mp

69-73 °C

mass shift

M+6

storage temp.

−20°C

SMILES string

COCC(=O)N(C(C)C(=O)OC)[13c]1[13c](C)[13cH][13cH][13cH][13c]1C

InChI

1S/C15H21NO4/c1-10-7-6-8-11(2)14(10)16(13(17)9-19-4)12(3)15(18)20-5/h6-8,12H,9H2,1-5H3/i6+1,7+1,8+1,10+1,11+1,14+1

InChI key

ZQEIXNIJLIKNTD-HEYPXIGASA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service