Skip to Content
Merck
All Photos(2)

Key Documents

748684

Sigma-Aldrich

Phosgene solution

15 wt. % in toluene

Synonym(s):

Carbon oxychloride, Carbonyl chloride, Phosgen

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
COCl2
CAS Number:
Molecular Weight:
98.92
Beilstein:
1098367
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

15 wt. % in toluene

density

0.924 g/mL at 25 °C

functional group

chloro

storage temp.

2-8°C

SMILES string

ClC(Cl)=O

InChI

1S/CCl2O/c2-1(3)4

InChI key

YGYAWVDWMABLBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phosgene is used as a reagent in organic synthesis for various reactions such as carbonylation reaction and synthesis of isocyanates. Phosgene is formed during the free radical induced degradation of chlorinated hydrocarbons. Determination of phosgene in air samples using thermal desorption (TD) followed by gas chromatography-mass spectrometry (GC-MS) has been reported.

Application

Phosgene solution (15 wt. % in toluene) has been used to synthesize the following peptide monomers:
  • γ-(4-propargyloxybenzyl)-L-glutamic acid N-carboxyanhydride (POB-L-Glu-NCA)
  • γ-(4,5-dimethoxy-2-nitrobenzyl)-L-glutamate N-carboxyanhydride (DMNB-L-Glu-NCA)
  • γ-(4,5-dimethoxy-2-nitrobenzyl)-D-glutamate N-carboxyanhydride (DMNB-D-Glu-NCA)
  • 2-nitrobenzyl(4-vinylphenyl)carbamate

Phosgene solution may be used in the synthesis of:
  • sulfonyl chlorides
  • Fmoc (Fluorenylmethyloxycarbonyl chloride)-taurylsulfonyl chloride
  • O-benzyl-L-serine carboxyanhydrides

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

UV?responsive degradable polymers derived from 1?(4?aminophenyl) ethane?1, 2?diol.
Ma L, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 53(9), 1161-1168 (2015)
Polypeptide vesicles with densely packed multilayer membranes.
Song Z, et al.
Soft Matter, 11(20), 4091-4098 (2015)
A kinetic study of the hydrolysis of phosgene in aqueous solution by pulse radiolysis.
Mertens R, et al.
Angewandte Chemie (International Edition in English), 33(12), 1259-1261 (1994)
The effect of side-chain functionality and hydrophobicity on the gene delivery capabilities of cationic helical polypeptides.
Zhang R, et al.
Biomaterials, 35(10), 3443-3454 (2014)
An Efficient Synthesis of N-Protected b-Amino-ethanesulfonylchlorides: Versatile Building Blocks for the Synthesis of Oligopeptidosulfonamides.
Brouwer AJ, et al.
Synthesis, 2000, 1579=84-1579=84 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service