Skip to Content
Merck
All Photos(1)

Key Documents

747106

Sigma-Aldrich

Difluoro{2-[(3,5-dimethyl-2H-pyrrol-2-ylidene-N)methyl]-3,5-dimethyl-1H-pyrrolato-N}boron

99% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15BF2N2
CAS Number:
Molecular Weight:
248.08
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99% (HPLC)

form

powder

mp

210-214 °C

SMILES string

CC(C=C1C)=C(/C=C2N=C(C)C=C\2C)N1B(F)F

InChI

1S/C13H15BF2N2/c1-8-5-10(3)17-12(8)7-13-9(2)6-11(4)18(13)14(15)16/h5-7H,1-4H3/b12-7-

InChI key

QZEVTPIWBHINIV-GHXNOFRVSA-N

Application

BODIPY laser dyes are used to generate fluorescent conjugates of proteins, nucleotides, oligonucleotides and dextrans, and to prepare fluorescent enzyme substrates, fatty acids, phospholipids, lipopolysaccharides, receptor ligands and polystyrene microspheres.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective fluorescent probes for live-cell monitoring of sulphide
Qian, Yong; Karpus, Jason; Kabil, Omer; et al.
Nature Communications, 2(Oct.), 1506/1-1506/7 (2011)
Siang Hui Lim et al.
Journal of medicinal chemistry, 53(7), 2865-2874 (2010-03-05)
To understand the effects of substitution patterns on photosensitizing the ability of boron dipyrromethene (BODIPY), two structural variations that either investigate the effectiveness of various iodinated derivatives to maximize the "heavy atom effect" or focus on the effect of extended
T Govender et al.
Bioresource technology, 114, 507-511 (2012-04-03)
In order to develop feasible production processes for microalgal biodiesel, the isolation of high neutral lipid producing microalgae is crucial. Since the established Nile Red (NR) method for detection of intracellular lipids has been successful only for some microalgae, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service