Skip to Content
Merck
All Photos(1)

Key Documents

740977

Sigma-Aldrich

L-Tyrosine-13C9,15N

endotoxin tested, 98 atom % 13C, 98 atom % 15N, 98% (CP)

Synonym(s):

L-4-Hydroxyphenylalanine-13C9,15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-HO-13C6H413CH213CH(15NH2)13CO2H
CAS Number:
Molecular Weight:
191.12
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % 13C
98 atom % 15N

Assay

98% (CP)

form

powder

mp

>300 °C (dec.) (lit.)

suitability

endotoxin tested

mass shift

M+10

SMILES string

[15NH2][13C@@H]([13CH2][13c]1[13cH][13cH][13c](O)[13cH][13cH]1)[13C](O)=O

InChI

1S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11H,5,10H2,(H,12,13)/t8-/m0/s1/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1

InChI key

OUYCCCASQSFEME-CMLFETTRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For R&D use only. Not for drug, household, or other uses. Any other use is the sole responsibility of the end user.

Endotoxin testing is done on the bulk form of the product before subdivision packaging. This bulk test does not guarantee that the subdivision or repackaged aliquot is endotoxin-free when it is received or used by the customer and does not imply suitability for any particular purpose. If the product must be endotoxin-free for the intended application, the product should be tested prior to actual use.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Min Xu et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1146, 122128-122128 (2020-05-04)
Ibotenic acid (IBA) is an amino acid and muscimol (MUS) is the decarboxyl derivative of IBA. They are mushroom neurotoxins with high polarity and low molecular weight. Only one transition (159->113 for IBA and 115->98 for MUS) can be found

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service