Skip to Content
Merck
All Photos(1)

Documents

739847

Sigma-Aldrich

Vitamin D2 (6,19,19-d3) solution

100 μg/mL in ethanol, 97 atom % D, 98% (CP)

Synonym(s):

Calciferol (6,19,19-d3) solution, Ercalciol (6,19,19-d3) solution, Ergocalciferol (6,19,19-d3) solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28D3H41O
Molecular Weight:
399.67
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.24

isotopic purity

97 atom % D

Quality Level

Assay

98% (CP)

form

solution

concentration

100 μg/mL in ethanol

technique(s)

mass spectrometry (MS): suitable

application(s)

detection

mass shift

M+3

storage temp.

−20°C

SMILES string

[2H]/C([2H])=C1CC[C@H](O)CC\1=C([2H])\C=C2/CCC[C@]3(C)[C@H](CC[C@@H]23)[C@H](C)\C=C\[C@H](C)C(C)C

InChI

1S/C28H44O/c1-19(2)20(3)9-10-22(5)26-15-16-27-23(8-7-17-28(26,27)6)12-13-24-18-25(29)14-11-21(24)4/h9-10,12-13,19-20,22,25-27,29H,4,7-8,11,14-18H2,1-3,5-6H3/b10-9+,23-12+,24-13-/t20-,22+,25-,26+,27-,28+/m0/s1/i4D2,13D

InChI key

MECHNRXZTMCUDQ-OQGZBSKRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Vitamin D2 is also known as ergocalciferol. It is produced by plants and mushrooms when exposed to sunlight.
Vitamin D2 (6,19,19-d3) is a deuterated vitamin D2 wherein C-6 and C-19 protons are replaced by deuterium.

Application

Vitamin D2 (6,19,19-d3) can be used as an analytical standard for the quantitative analysis of vitamin D2 concentration in human blood via LC-MS/MS.

Packaging

1 mL in ampule

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F

Flash Point(C)

14 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sterol and vitamin D2 concentrations in cultivated and wild grown mushrooms: Effects of UV irradiation
Teichmann A, et al.
LWT--Food Science and Technology, 40(5), 815-822 (2007)
Application of a new vitamin D blood test on the Emirati population
Al-Dabbagh, B et al.
The Journal of Steroid Biochemistry and Molecular Biology, 180(5), 118-128 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service