Skip to Content
Merck
All Photos(3)

Key Documents

737925

Sigma-Aldrich

Decylphosphonic acid

97%

Synonym(s):

n-Decylphosphonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H23O3P
CAS Number:
Molecular Weight:
222.26
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

solid

mp

96-101 °C

SMILES string

CCCCCCCCCCP(O)(O)=O

InChI

1S/C10H23O3P/c1-2-3-4-5-6-7-8-9-10-14(11,12)13/h2-10H2,1H3,(H2,11,12,13)

InChI key

DZQISOJKASMITI-UHFFFAOYSA-N

General description

Decylphosphonic acid(DPA) is a phosphono-alkane derivative that forms a self-assembled monolayer(SAM) that can be used as an interfacial modifier.

Application

DPA doped polyaniline(PANI) can be used as a protective coating against corrosion. It can also form a doublelayer thin film with the dye solution which may be assembled on a counter electrode for the fabrication of dye sensitized solar cells(DSSCs).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance nanostructured inorganic- organic heterojunction solar cells
Chang JA, et al.
Nano Letters, 10(7), 2609-2612 (2010)
Soundarrajan Chandrasekaran et al.
Chemical science, 10(16), 4469-4475 (2019-05-07)
Hydrogen production through direct sunlight-driven water splitting in photo-electrochemical cells (PECs) is a promising solution for energy sourcing. PECs need to fulfill three criteria: sustainability, cost-effectiveness and stability. Here we report an efficient and stable photocathode platform for H2 evolution
Kinetics of self-assembled layer formation on iron
Felhhosi I, et al.
Electrochimica Acta, 47(13-14), 2335-2340 (2002)
Preparation and characterization of polyaniline in reversed micelles of decylphosphonic acid for active corrosion protection coatings
Perrin FX, et al.
European Polymer Journal, 66(7), 253-265 (2015)
Daibin Kuang et al.
Journal of the American Chemical Society, 128(12), 4146-4154 (2006-03-23)
Two novel heteroleptic sensitizers, Ru((4,4-dicarboxylic acid-2,2'-bipyridine)(4,4'-bis(p-hexyloxystyryl)-2,2-bipyridine)(NCS)2 and Ru((4,4-dicarboxylic acid-2,2'-bipyridine)(4,4'-bis(p-methoxystyryl)-2,2'-bipyridine) (NCS)2, coded as K-19 and K-73, respectively, have been synthesized and characterized by 1H NMR, FTIR, UV-vis absorption, and emission spectroscopy and excited-state lifetime and spectroelectrochemical measurements. The introduction of the

Articles

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service