733733
D-Mannose-4-13C
99 atom % 13C, 97% (CP)
Synonym(s):
D-Mannopyranose-4-13C
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
13CC5H12O6
Molecular Weight:
181.15
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Recommended Products
isotopic purity
99 atom % 13C
Assay
97% (CP)
form
solid
mp
133-140 °C (lit.)
mass shift
M+1
SMILES string
OC[C@H]1OC(O)[C@@H](O)[C@@H](O)[13C@@H]1O
InChI
1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5+,6?/m1/s1/i3+1
InChI key
WQZGKKKJIJFFOK-XEIQMVQRSA-N
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service