Skip to Content
Merck
All Photos(1)

Key Documents

73244

Sigma-Aldrich

1-Hexyl-3-methylimidazolium tetrafluoroborate

≥97.0% (HPLC)

Synonym(s):

HMIMBF4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19BF4N2
CAS Number:
Molecular Weight:
254.08
Beilstein:
8366236
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥97.0% (HPLC)

form

liquid

impurities

≤1.0% water

refractive index

n20/D 1.430

density

1.149 g/mL at 20 °C (lit.)

SMILES string

F[B-](F)(F)F.CCCCCCn1cc[n+](C)c1

InChI

1S/C10H19N2.BF4/c1-3-4-5-6-7-12-9-8-11(2)10-12;2-1(3,4)5/h8-10H,3-7H2,1-2H3;/q+1;-1

InChI key

MFXLOVLEQJRXFP-UHFFFAOYSA-N

Other Notes

Enzyme catalysis in ionic liquids[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S.H. Schofer et al.
Chemical Communications (Cambridge, England), 425-425 (2001)
E Diaz et al.
Ecotoxicology and environmental safety, 162, 29-34 (2018-07-01)
The ecotoxicity and inhibition of 12 imidazolium ionic liquids (ILs) with alkyl chain from C4 to C10 and chloride (Cl-), tetrafluoroborate (BF4-) and bis(trifluoromethanesulfonyl)imide (NTf2-) anions have been studied by means of respiration inhibition assays using activated sludge collected from
Kunze Du et al.
Food chemistry, 244, 190-196 (2017-11-10)
A simple and green ionic liquid-based vortex-forced matrix solid phase dispersion (IL-VFMSPD) method was presented to simultaneously extract 5-hydroxymethyl furfurol (5-HMF) and iridoid glycosides in Fructus Corni by ultra-high performance liquid chromatography. Ionic liquid was used as a green elution
Renata Kołodziejska et al.
Chirality, 30(2), 206-214 (2017-11-16)
The application of ionic liquids as solvents for transesterification of prochiral pirymidine acyclonucleoside using lipase (EC 3.1.1.3) Amano PS from Burkholderia cepacia (BCL) is reported. The effect of using medium reaction, acyl group donor, and temperature on the activity and
Sylwia Bajkacz et al.
Molecules (Basel, Switzerland), 25(14) (2020-07-18)
This work aimed to comprehensively evaluate the potential and effectiveness of deep eutectic solvents (DESs) in the extraction of seven catechins from various tea samples. Different combinations of DES were used, consisting of Girard's reagent T (GrT) in various mixing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service