Skip to Content
Merck
All Photos(1)

Key Documents

727695

Sigma-Aldrich

1-Allyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide

≥98.5% (HPLC)

Synonym(s):

AMIMTFSI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11F6N3O4S2
CAS Number:
Molecular Weight:
403.32
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.5% (HPLC)

form

liquid

impurities

≤1% water

functional group

fluoro

SMILES string

C[n+]1ccn(CC=C)c1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

InChI

1S/C7H11N2.C2F6NO4S2/c1-3-4-9-6-5-8(2)7-9;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h3,5-7H,1,4H2,2H3;/q+1;-1

InChI key

DHMWATGUEVQTIY-UHFFFAOYSA-N

General description

1-Allyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide is an ionic liquid that can be prepared by reacting 1-allyl-3-methylimidazolium chloride and lithium bis(trifluoromethylsulfonyl)imide under microwave conditions.

Application

1-Allyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide can be used to prepare AMIMTFSI-propylene carbonate based electrolyte, with potential application in lithium-ion batteries. It also finds application in alkanes/alkenes separation process. AMIMTFSI can be used as a surface protector to develop luminescent silicon nanoparticles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermodynamics and selectivity of separation based on activity coefficients at infinite dilution of various solutes in 1-allyl-3-methylimidazolium bis {(trifluoromethyl) sulfonyl} imide ionic liquid.
Wlazlo M, et al.
The Journal of Chemical Thermodynamics, 102, 39-47 (2016)
(Liquid+ liquid) equilibrium for the ternary systems {heptane+ toluene+ 1-allyl-3-methylimidazolium bis (trifluoromethylsulfonyl) imide} and {heptane+ toluene+ 1-methyl-3-propylimidazolium bis (trifluoromethylsulfonyl) imide} ionic liquids
Garcia, Silvia, et al.
The Journal of Chemical Thermodynamics, 43.11, 1641-1645 (2011)
Hassan M Albishri et al.
Talanta, 200, 107-114 (2019-05-01)
Acetylcholine (ACh) and its precursor choline (Ch) play important roles in many biological processes. It is expected that Alzheimer's disease is occurred due to the reduction in synthesis of ACh. On the other hand, the increase in the level of
Liquid?liquid equilibrium data for {heptane+ aromatic+ 1-(2-hydroxyethyl)-3-methylimidazolium bis (trifluoromethylsulfonyl) imide ([hemim][NTf2])} ternary systems
Ebrahimi, M., et al.
Journal of Chemical and Engineering Data, 59.2, 197-204 (2014)
Synthesis and properties of pyrrolidinium and piperidinium bis (trifluoromethanesulfonyl) imide ionic liquids with allyl substituents
Yim, Tae Eun, et al.
J. Korean Chem. Soc., 28.9, 1567-1567 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service