Skip to Content
Merck
All Photos(1)

Documents

727075

Sigma-Aldrich

(R)-(−)-Mandelic acid

ChiPros®, produced by BASF, 99%

Synonym(s):

(R)-α-Hydroxyphenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein:
2691094
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

grade

produced by BASF

Quality Level

Assay

≥98.5%
99%

form

crystals

optical purity

enantiomeric excess: ≥98.5%

mp

131-133 °C (lit.)

SMILES string

O[C@@H](C(O)=O)c1ccccc1

InChI

1S/C8H8O3/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7,9H,(H,10,11)/t7-/m1/s1

InChI key

IWYDHOAUDWTVEP-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(R)-(−)-Mandelic acid is a chiral resolving agent, which is also used as a chiral building block in the preparation of semisynthetic cephalosporins, antitumor agents, and penicillins.

Application

(R)-(−)-Mandelic acid can be used:
  • As a chiral acid in the separation of diastereomeric salts for the synthesis of chiral pyridoindole based building block.
  • In the study of chiral acid selectivity of poly(3,4-ethylenedioxythiophene) (PEDOT) based chiral conducting polymers.

Legal Information

ChiPros is a registered trademark of BASF SE

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

>374.0 °F

Flash Point(C)

> 190 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiral acid selectivity displayed by PEDOT electropolymerised in the presence of chiral molecules
Sulaiman Y and Kataky R
Analyst, 137(10), 2386-2393 (2012)
A Short Synthesis of the 2-Bromo-N, 9-dimethyl-6, 7, 8, 9-tetrahydro-5 H-pyrido [2, 3-b] indol-6-amine Building Block
Sreenivasachary N, et al.
Organic Process Research & Development, 23(11), 2521-2526 (2019)
Biocatalytic synthesis of (R)-(−)-mandelic acid from racemic mandelonitrile by a newly isolated nitrilase-producer Alcaligenes sp. ECU0401
He YC, et al.
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao, 18(6), 677-680 (2007)
M Monier et al.
International journal of biological macromolecules, 55, 207-213 (2013-01-30)
An enantioselective S-mandelic acid (S-MA) imprinted chitosan (SMIC) was prepared by cross-linking of chitosan using formaldehyde cross-linker, in the presence of S-MA as an imprint template molecule and 0.5% acetic acid solution as a solvent. Non-imprinted cross-linked chitosan (NIC) as
Chao Ma et al.
Chirality, 23(5), 379-382 (2011-04-14)
This work reports the chiral separation of D,L-mandelic acid with cellulose membranes. Cellulose was chosen as membrane material because it possesses multichiral carbon atoms in its molecular structure unit. The flux and permselective properties of membrane using aqueous solutions of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service