Skip to Content
Merck
All Photos(5)

Key Documents

725110

Sigma-Aldrich

Lithium manganese nickel oxide

greener alternative

spinel, powder, <0.5 μm particle size (BET), >99%

Synonym(s):

LMNO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li2Mn3NiO8
CAS Number:
Molecular Weight:
365.38
MDL number:
UNSPSC Code:
26111700
NACRES:
NA.23

grade

battery grade

Assay

>99%

form

powder

mol wt

Mw 365.38 g/mol

composition

Li2Mn3NiO8

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

<0.5 μm (BET)

mp

>290 °C

application(s)

battery manufacturing

greener alternative category

General description

Lithium manganese nickel oxide (LMNO) is a class of electrode material that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Features and Benefits

Cathode Materials for High Energy Density Li Ion Rechargeable Batteries; sub-micron particle size results in increased surface area of electrodes. The composition listed allows the preparation of battery electrodes with enhanced performance and durability.

Li-Batteries, material for electrodes design and manufacturing

Legal Information

Product of Engi-Mat Co.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kuthanapillil M Shaju et al.
Dalton transactions (Cambridge, England : 2003), (40), 5471-5475 (2008-12-17)
Disordered and ordered forms of nano-Li[Ni(0.5)Mn(1.5)]O(4) spinel, have been prepared by a one-pot resorcinol-formaldehyde synthesis. Lithium intercalation into disordered nano-Li[Ni(0.5)Mn(1.5)]O(4-delta) reveals good rate capability and cycling stability. It delivers 95.5% of the capacity at a rate of 10C (1500 mA
Wu, H. M.; Belharouak, I.; Deng, H.;
Journal of the Electrochemical Society, 156, A1047-A1047 (2009)
Hagh, N. Marandian; Amatucci, G. G.
Electrochemical Society Transactions, 11 (29), 21-26 (2008)
Electrodes with high power and high capacity for rechargeable lithium batteries
Kang K, et al.
Science, 311(5763), 977-980 (2006)
Towards greener and more sustainable batteries for electrical energy storage
Larcher D and Tarascon J
Nature Chemistry, 7(1), 19-19 (2015)

Articles

Nanomaterials for Energy Storage in Lithium-ion Battery Applications

HEVs address rising fuel costs and emissions concerns, utilizing battery packs alongside internal combustion engines for enhanced performance.

HEVs address rising fuel costs and emissions concerns, utilizing battery packs alongside internal combustion engines for enhanced performance.

HEVs address rising fuel costs and emissions concerns, utilizing battery packs alongside internal combustion engines for enhanced performance.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service