Skip to Content
Merck
All Photos(3)

Documents

715700

Sigma-Aldrich

3-Furonitrile

97%

Synonym(s):

Furan-3-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3NO
CAS Number:
Molecular Weight:
93.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

refractive index

n20/D 1.471

mp

28-32 °C

density

1.073 g/mL at 25 °C

SMILES string

N#Cc1ccoc1

InChI

1S/C5H3NO/c6-3-5-1-2-7-4-5/h1-2,4H

InChI key

ZQFYJHMUAWCEBH-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

120.0 °F - closed cup

Flash Point(C)

48.9 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service