Skip to Content
Merck
All Photos(1)

Key Documents

709727

Sigma-Aldrich

3-(Trimethylsilyl)ethynylbenzaldehyde

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14OSi
CAS Number:
Molecular Weight:
202.32
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.547

density

0.962 g/mL at 25 °C

functional group

aldehyde

storage temp.

2-8°C

SMILES string

C[Si](C)(C)C#Cc1cccc(C=O)c1

InChI

1S/C12H14OSi/c1-14(2,3)8-7-11-5-4-6-12(9-11)10-13/h4-6,9-10H,1-3H3

InChI key

UPZYBRHQVJTPBO-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aoibhín A Cullen et al.
Frontiers in chemistry, 8, 584060-584060 (2020-11-17)
A novel 4,4-difuoro-4-bora-3a,4a-diaza-s-indacene (BODIPY) copolymer with diethynylbenzene has been synthesised, and its ability to act as a photosensitiser for the photocatalytic generation of hydrogen was investigated by time-resolved spectroscopic techniques spanning the ps- to ns-timescales. Both transient absorption and time-resolved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service