Skip to Content
Merck
All Photos(2)

Key Documents

705055

Sigma-Aldrich

4,4′-Azopyridine

Synonym(s):

Azobis(4-pyridine)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8N4
CAS Number:
Molecular Weight:
184.20
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

96-101 °C

SMILES string

c1cc(ccn1)N=Nc2ccncc2

InChI

1S/C10H8N4/c1-5-11-6-2-9(1)13-14-10-3-7-12-8-4-10/h1-8H

InChI key

XUPMSLUFFIXCDA-UHFFFAOYSA-N

General description

4,4′-Azopyridine is an aromatic heterocyclic compound used as a ligand in the formation of [HgI2(4,4′-azopyridine)]n complex.

Application

4,4′-Azopyridine can be used:
  • To prepare porous coordination polymers (PCPs) by reacting with Zn(NO3)2 and 1,4-benzenedicarboxylic acid.
  • As a reagent for the conversion of aliphatic alcohols into disulfides under Mitsunobu conditions.
  • To prepare 4,4′-azopyridine-bridged binuclear zinc(II) complexes.

Reactant for preparation of:
  • Flexible porous coordination polymers constructed from 1,2-bis(4-pyridyl)hydrazine via solvothermal in situ reduction reaction
  • Metal-organic frameworks based on transition-metal carboxylate clusters as secondary building units
  • Heteroaromatic azo compounds under Mitsunobu conditions
  • MnII, CoII, and ZnII coordination polymers
  • Low-dimensional and porous coordination compounds capable of supramolecular aromatic interactions

Reagent in:
  • Facile Mitsunobu esterification reactions

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Flexible porous coordination polymers constructed from 1, 2-bis (4-pyridyl) hydrazine via solvothermal in situ reduction of 4, 4′-azopyridine
Liu X, et al.
Dalton Transactions, 40(34), 8549-8554 (2011)
A novel HgI2 adduct with an azopyridine ligand: synthesis, structure and optical refractive effect of [HgI2 (4, 4?-azopyridine)] n
Niu, Yunyin and Song
CrystEngComm, 3, 152-154 (2001)
4, 4?-Azopyridine as an easily prepared and recyclable oxidant for synthesis of symmetrical disulfides from thiols or alkyl halides (tosylates)/thiourea}
Khalili, Dariush and Iranpoor
Journal of Sulfur Chemistry, 36, 544-555 (2015)
Heteroaromatic azo compounds as efficient and recyclable reagents for direct conversion of aliphatic alcohols into symmetrical disulfides
Iranpoor N, et al.
Tetrahedron Letters, 53(51), 6913-6915 (2012)
A novel 4, 4′-azopyridine-bridged binuclear zinc complex
Zhu L, et al.
Journal of Coordination Chemistry, 56(17), 1447-1453 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service