Skip to Content
Merck
All Photos(2)

Documents

703184

Sigma-Aldrich

Indium tin oxide coated glass slide, square

surface resistivity 30-60 Ω/sq, slide

Synonym(s):

ITO, ITO coated slide, square

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
In2O3 · (SnO2)x
CAS Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

surface resistivity

30-60 Ω/sq

L × W × thickness

25 mm × 25 mm × 1.1 mm

transmittance

84% (nominal at 550nm)

refractive index

n20/D 1.517

SMILES string

O=[Sn]=O.O=[In]O[In]=O

InChI

1S/2In.5O.Sn

InChI key

LNNWKAUHKIHCKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Substrates

Substrate: unpolished float glass, Si02 passivated (200-300 Å)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Plexcore® organic conductive inks are electronic grade inks formulated for use in the hole injection layers of OLEDs.

Plexcore® organic conductive inks are electronic grade inks formulated for use in the hole injection layers of OLEDs.

Plexcore® organic conductive inks are electronic grade inks formulated for use in the hole injection layers of OLEDs.

Plexcore® organic conductive inks are electronic grade inks formulated for use in the hole injection layers of OLEDs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service