Skip to Content
Merck
All Photos(1)

Key Documents

69893

Sigma-Aldrich

2-Morpholinoethyl isocyanide

≥98.0% (GC)

Synonym(s):

4-(2-Isocyanoethyl)morpholine, MEI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12N2O
CAS Number:
Molecular Weight:
140.18
Beilstein:
4666990
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.469

bp

72-73 °C/0.7 mmHg (lit.)

density

1.017 g/mL at 20 °C (lit.)

functional group

amine
ether
isonitrile

storage temp.

−20°C

SMILES string

[C-]#[N+]CCN1CCOCC1

InChI

1S/C7H12N2O/c1-8-2-3-9-4-6-10-7-5-9/h2-7H2

InChI key

MFRZPLYKVDHOSN-UHFFFAOYSA-N

Application

2-Morpholinoethyl isocyanide can be used for the synthesis of:
  • Nitrogen acyclic carbene (NAC) complexes to be used as catalysts for the synthesis of phenol and hydration of alkynes.
  • Variety of imidazo[1,2-a]pyridines and α-acylaminoamides by Ugi condensation reaction.
  • 2-Aminobenzoxazoles and 3-aminobenzoxazines having important therapeutic activities.

It can also be used as a ligand in bis-cyclometalated iridium complexes for the organic light emitting diodes (OLEDs).

Other Notes

Potent coupling reagent for peptide synthesis giving higher yields and less racemization than DCCI; Reagent used in a Passerini-like reaction to alpha-hydroxy amides

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael J Kinney et al.
PloS one, 12(11), e0188660-e0188660 (2017-11-29)
Horizontal behavior of highly migratory marine species is difficult to decipher because animals are wide-ranging, spend minimal time at the ocean surface, and utilize remote habitats. Satellite telemetry enables researchers to track individual movements, but population level inferences are rare
Microwave-assisted solid-phase Ugi four-component condensations.
Hoel AML and Nielsen J
Tetrahedron Letters, 40(20), 3941-3944 (1999)
David Jakubosky et al.
Nature communications, 11(1), 2927-2927 (2020-06-12)
Structural variants (SVs) and short tandem repeats (STRs) comprise a broad group of diverse DNA variants which vastly differ in their sizes and distributions across the genome. Here, we identify genomic features of SV classes and STRs that are associated
Karen A Bjorndal et al.
Global change biology, 23(11), 4556-4568 (2017-04-06)
Somatic growth is an integrated, individual-based response to environmental conditions, especially in ectotherms. Growth dynamics of large, mobile animals are particularly useful as bio-indicators of environmental change at regional scales. We assembled growth rate data from throughout the West Atlantic
New and Easily Accessible Nitrogen Acyclic Gold (I) Carbenes: Structure and Application in the Gold-Catalyzed Phenol Synthesis as well as the Hydration of Alkynes.
Hashmi ASK, et al.
Advanced Synthesis & Catalysis, 352(8), 1315-1337 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service