Skip to Content
Merck
All Photos(2)

Documents

696528

Sigma-Aldrich

3-Methyl-2-butenylboronic acid pinacol ester

96%

Synonym(s):

2-(3-Methyl-but-2-enyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,3-Dimethylallylboronic acid pinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H21BO2
CAS Number:
Molecular Weight:
196.09
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.4395

bp

42-45 °C/0.6 mmHg

density

0.885 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C\C(C)=C\CB1OC(C)(C)C(C)(C)O1

InChI

1S/C11H21BO2/c1-9(2)7-8-12-13-10(3,4)11(5,6)14-12/h7H,8H2,1-6H3

InChI key

QXDMQRNIDKVRCN-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.1 °F

Flash Point(C)

76.7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service