Skip to Content
Merck
All Photos(1)

Key Documents

68742

Sigma-Aldrich

Methyl phenyl sulfone

≥98.0% (GC)

Synonym(s):

(Methylsulfonyl)benzene, (Phenylsulfonyl)methane, Phenyl methyl sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO2C6H5
CAS Number:
Molecular Weight:
156.20
Beilstein:
1906914
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (GC)

form

crystals

mp

85-87 °C

functional group

sulfone

SMILES string

CS(=O)(=O)c1ccccc1

InChI

1S/C7H8O2S/c1-10(8,9)7-5-3-2-4-6-7/h2-6H,1H3

InChI key

JCDWETOKTFWTHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Versatile building block for the construction of diverse functionalities[1][2][3][4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M.C. Mussatto et al.
Journal of the Chemical Society. Perkin Transactions 1, 260-260 (1980)
P.A. Bartlett et al.
Journal of the American Chemical Society, 100, 4852-4852 (1978)
Shaofang Sun et al.
Water research, 156, 1-8 (2019-03-22)
Very recently, several studies have found that homogeneous reducing agents (e.g., sodium thiosulfate (Na2S2O3), and sodium sulfite (Na2SO3)) can activate ferrate to enhance the degradation of selected contaminants. In this work, it was found that heterogeneous carbon nanotube (CNT) could
M. Julia et al.
Tetrahedron Letters, 277-277 (1976)
Dandan Rao et al.
Water research, 188, 116481-116481 (2020-10-12)
Although permanganate activation by sodium sulfite (Mn(VII)/Na2SO3) has shown great potential for rapid abatement of organic contaminants, the limited reactivity under alkaline conditions and undesirable Mn residual may prevent its widespread application. To solve these challenges, calcium sulfite (CaSO3) was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service