Skip to Content
Merck
All Photos(1)

Key Documents

686719

Sigma-Aldrich

2-Fluoro-6-methylbenzaldehyde

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7FO
CAS Number:
Molecular Weight:
138.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
96%

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.525

density

1.151 g/mL at 20 °C

functional group

aldehyde
fluoro

storage temp.

2-8°C

SMILES string

Cc1cccc(F)c1C=O

InChI

1S/C8H7FO/c1-6-3-2-4-8(9)7(6)5-10/h2-5H,1H3

InChI key

BGQBJWPZBBMKEI-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service