Skip to Content
Merck
All Photos(3)

Documents

684589

Sigma-Aldrich

2-Methoxy-3-pyridinylboronic acid

≥95.0%

Synonym(s):

2-Methoxy-3-pyridineboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8BNO3
CAS Number:
Molecular Weight:
152.94
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0%

form

solid

mp

140-144 °C

storage temp.

2-8°C

SMILES string

COc1ncccc1B(O)O

InChI

1S/C6H8BNO3/c1-11-6-5(7(9)10)3-2-4-8-6/h2-4,9-10H,1H3

InChI key

NVOLYUXUHWBCRJ-UHFFFAOYSA-N

General description

May contain varying amounts of anhydride.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service