Skip to Content
Merck
All Photos(2)

Key Documents

675067

Sigma-Aldrich

12-Mercaptododecanoic acid

96%

Synonym(s):

MDA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H24O2S
CAS Number:
Molecular Weight:
232.38
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

96%

form

solid

mp

45-50 °C

storage temp.

2-8°C

SMILES string

OC(=O)CCCCCCCCCCCS

InChI

1S/C12H24O2S/c13-12(14)10-8-6-4-2-1-3-5-7-9-11-15/h15H,1-11H2,(H,13,14)

InChI key

SDAWVOFJSUUKMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

12-Mercaptododecanoic acid (MDA) is a carboxylic acid terminated thiol that forms self-assembled monolayers (SAMs) on a variety of surfaces. It provides a blocking effect and immobilizes the surface atoms.

Application

MDA can functionalize gold nanoparticles which can form screen printed electrodes for electrochemical detection. It can also form ligands with cadmium selenium (CdSe) quantum dots (ODs) which can be deposited on titanium oxide(TiO2) based films.
This compound is used to produce hydrophilic SAMs. The resulting monolayers which are terminated with carboxylic acids can be further functionalized to introduce more complex end groups. Used in a study of the effect of molecular structure on the precision of copper-bound α,ω-mercaptoalkanoic acid "molecular ruler" stacks.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Langmuir, 23, 638-638 (2000)
Development of a label-free impedance biosensor for detection of antibody-antigen interactions based on a novel conductive linker.
Chen C, et al.
Biosensors And Bioelectronics, 26(6), 3072-3076 (2011)
Aminoalkanoic Acids as Alternatives to Mercaptoalkanoic Acids for the Linker-Assisted Attachment of Quantum Dots to TiO2.
Rivera-Gonza?lez N, et al.
Langmuir, 32(36), 9206-9215 (2016)
Synthesis and assembly of gold and iron oxide particles within an emulsion droplet; facile production of Core@ Shell particles.
Sachdev S, et al.
Colloids and Interface Science Communications, 16(6), 14-18 (2017)
Iron oxide/gold core/shell nanomagnetic probes and CdS biolabels for amplified electrochemical immunosensing of Salmonella typhimurium.
Freitas M, et al.
Biosensors And Bioelectronics, 51(6), 195-200 (2014)

Articles

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service