Skip to Content
Merck
All Photos(1)

Documents

658758

Sigma-Aldrich

1H,1H,2H,2H-Perfluorodecyltriethoxysilane

97%

Synonym(s):

Triethoxy-1H,1H,2H,2H-perfluorodecylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19F17O3Si
CAS Number:
Molecular Weight:
610.38
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

refractive index

n20/D 1.342

bp

209-230 °C

density

1.389 g/mL at 25 °C

SMILES string

CCO[Si](CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)(OCC)OCC

InChI

1S/C16H19F17O3Si/c1-4-34-37(35-5-2,36-6-3)8-7-9(17,18)10(19,20)11(21,22)12(23,24)13(25,26)14(27,28)15(29,30)16(31,32)33/h4-8H2,1-3H3

InChI key

MLXDKRSDUJLNAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1H,1H,2H,2H-Perfluorodecyltriethoxysilane(PFDTES) is a fluoroalkylsilanes(FAS) with low surface energy and high wettability due to the presence of fluorinated carbons. It is used as a superhydrophobic agent for synthesizing various functional coatings.

Application

It was used to functionalize hydrophobic ceramic membranes with nanosized pores. It was also used to functionalize Cu2(OH)3NO3 crystal  to render it hydrophobic. The product may be used to coat nanosized medical devices.
PFDTES is used for the superhydrophobicity of polyvinylidene fluoride(PVDF) to protect the metal surfaces from fouling. A cost effective and scalable superamphiphobic spray coating of PFDTES on stainless steel surfaces may potentially be used to create anti-icing and stain-resistant surfaces. It may be used to modify the surface of poly(methyl methacrylate(PMMA) for the fabrication of microfluidic chips.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Desalination of sodium chloride solutions and seawater with hydrophobic ceramic membranes
Gazagnes L, et al.
Desalination, 217, 260-266 (2007)
Grafting of ZrO2 powder and ZrO2 membrane by fluoroalkylsilanes.
Krajewski SR, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 243(1-3), 43-47 (2004)
New approach to fabricate an extremely super-amphiphobic surface based on fluorinated silica nanoparticles.
Sheen Y, et al.
Journal of Polymer Science. Part B, Polymer Physics, 46(18), 1984-1990 (2008)
Surface modification of silicon and polydimethylsiloxane surfaces with vapor-phase-deposited ultrathin fluorosilane films for biomedical nanodevices.
Bhushan B, et al.
J. Vac. Sci. Technol., 24(4), 1197-1202 (2006)
Jacopo Bernardini et al.
ACS applied materials & interfaces, 12(15), 18046-18055 (2020-03-20)
The interaction of rising gas bubbles with submerged air-repelling or air-attracting surfaces is relevant to various technological applications that rely on gas-microvolume handling or removal. This work demonstrates how submerged metal meshes with super air-attracting/repelling properties can be employed to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service