Skip to Content
Merck
All Photos(1)

Documents

650137

Sigma-Aldrich

Benzofurazan

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4N2O
CAS Number:
Molecular Weight:
120.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

bp

75-85 °C/20 mmHg (lit.)

mp

47-51 °C (lit.)

SMILES string

c1ccc2nonc2c1

InChI

1S/C6H4N2O/c1-2-4-6-5(3-1)7-9-8-6/h1-4H

InChI key

AWBOSXFRPFZLOP-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomofumi Santa et al.
Biomedical chromatography : BMC, 21(11), 1207-1213 (2007-06-26)
The applicability of benzofurazan derivatization regents to carboxylic acids analysis in LC/ESI-MS/MS (high-performance liquid chromatography/electrospray ionization tandem mass spectrometry) was examined. The product ion spectra of DAABD-AE {4-[2-(N,N-dimethylamino)ethylaminosulfonyl]-7-(2-aminoethylamino)-2,1,3-benzoxadiazole}, DAABD-PZ {4-[2-(N,N-dimethylamino)ethylaminosulfonyl]-7-N-piperazino-2,1,3-benzoxadiazole}, DAABD-PiCZ {4-[4-carbazoylpiperidin-1-yl]-7-[2-(N,N-dimethylamino)ethylaminosulfonyl]-2,1,3-benzoxadiazole}, DAABD-ProCZ {4-[2-carbazoylpyrrolidin-1-yl]-7-[2-(N,N-dimethylamino) ethylaminosulfonyl]-2,1,3-benzoxadiazole} and DAABD-Apy {4-[2-(N,N-dimethylamino)ethylaminosulfonyl]-7-(3-aminopyrrolidin-1-yl)-2,1,3-benzoxadiazole}, and their
Zhipeng Liu et al.
The Journal of organic chemistry, 76(24), 10286-10290 (2011-11-18)
A dicyanovinyl-substituted benzofurazan derivative (C1) was prepared as an efficient ratiometric chemosensor for cyanide anion detection in aqueous acetonitrile solution. Mechanism studies suggested that the nucleophilic addition of cyanide to the α-position of the dicyanovinyl group blocked the ICT progress
Zhenghao Yang et al.
Dalton transactions (Cambridge, England : 2003), 40(10), 2173-2176 (2010-11-23)
Terpyridine/benzofurazan conjugation results in a new hybrid fluorophore of the colorimetric sensing ability for Fe(2+) and fluorimetric sensing ability for XII group cations. The improved emission properties and cell imaging ability imply it is a suitable platform to construct a
Alex Brown et al.
The journal of physical chemistry. A, 116(1), 46-54 (2011-12-06)
General chemical strategies which provide controlled changes in the emission or absorption properties of biologically compatible fluorophores remain elusive. One strategy employed is the conversion of a fluorophore-attached alkyne (or azide) to a triazole through a copper-catalyzed azide-alkyne coupling (CuAAC)
Maki Onoda et al.
Chemical communications (Cambridge, England), (14)(14), 1848-1850 (2005-03-30)
Strong fluorescence signals were observed after the reaction of novel reagents with hydroperoxides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service