645524
Silica gel Inorganic Sorbent
technical grade, 200-425 mesh
About This Item
Recommended Products
Product Name
Silica gel, technical grade, pore size 60 Å, 200-425 mesh particle size
grade
technical grade
Quality Level
description
Residual water (≤12%)
form
powder
manufacturer/tradename
Sigma-Aldrich
technique(s)
LPLC: suitable
surface area
500-600 m2/g , approx.
matrix
Silica
matrix active group
silica
particle size
200-425 mesh
35-75 μm
pore size
0.75 cm3/g pore volume (approx.)
60 Å
pH
6.5-7.5
bp
2230 °C
mp
>1600 °C
separation technique
hydrophilic interaction (HILIC)
SMILES string
O=[Si]=O
InChI
1S/O2Si/c1-3-2
InChI key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | 717185 | 717177 | 227196 |
---|---|---|---|
separation technique hydrophilic interaction (HILIC) | separation technique hydrophilic interaction (HILIC) | separation technique hydrophilic interaction (HILIC) | separation technique hydrophilic interaction (HILIC) |
matrix active group silica | matrix active group silica | matrix active group silica | matrix active group silica |
pore size 0.75 cm3/g pore volume (approx.), 60 Å | pore size 0.7-0.85 cm3/g pore volume (approx.), 60 Å | pore size 0.7-0.85 cm3/g pore volume (approx.), 60 Å | pore size ~0.8 cm3/g pore volume, 60 Å |
matrix Silica | matrix Silica | matrix Silica | matrix Silica |
technique(s) LPLC: suitable | technique(s) LPLC: suitable | technique(s) LPLC: suitable | technique(s) LPLC: suitable |
Application
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service