Skip to Content
Merck
All Photos(1)

Documents

644285

Sigma-Aldrich

[2-[2-(2-Methoxyethoxy)ethoxy]ethoxy]p-toluenesulfonate

90%

Synonym(s):

Tri(ethylene glycol) monomethyl ether tosylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22O6S
CAS Number:
Molecular Weight:
318.39
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

90%

SMILES string

COCCOCCOCCOS(=O)(=O)c1ccc(C)cc1

InChI

1S/C14H22O6S/c1-13-3-5-14(6-4-13)21(15,16)20-12-11-19-10-9-18-8-7-17-2/h3-6H,7-12H2,1-2H3

InChI key

IUDNRKGPFWUYIC-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service