634506
Mercsorb® mercury spill kit
For one-time use only
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
47131905
NACRES:
NB.35
Recommended Products
Related Categories
General description
Kit is designed to handle all stages of cleaning up mercury spills. Does not contain any chemicals designed to make the mercury less toxic or less dangerous. For one-time use only.
Contents:
Contents:
- Amalgamation powder
- Indicator powder
- Vapor suppressor, shaker bottle
- Aspirator bottle
- Waste collection bottle
- Mixing tub with spatula
- Chemical sponge
- Nitrile gloves
- Disposal bag and twist tie
- Wisk on pan
Legal Information
Mercsorb is a registered trademark of NPS Corp.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service