Skip to Content
Merck
All Photos(3)

Key Documents

632961

Sigma-Aldrich

5′-Hexyl-2,2′-bithiophene-5-boronic acid pinacol ester

97%

Synonym(s):

2-(5′-Hexyl-2,2′-bithien-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-Tetramethyl-2-[5′-hexyl-2,2′-bithien-5-yl]-1,3,2-dioxaborolane, 5′-N-Hexyl-2,2′-bithiophene-5-boronic acid pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5′-N-hexyl-2,2′-bithiophene, 5-Hexyl-5′-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,2′-bithiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H29BO2S2
CAS Number:
Molecular Weight:
376.38
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

36-40 °C (lit.)

SMILES string

CCCCCCc1ccc(s1)-c2ccc(s2)B3OC(C)(C)C(C)(C)O3

InChI

1S/C20H29BO2S2/c1-6-7-8-9-10-15-11-12-16(24-15)17-13-14-18(25-17)21-22-19(2,3)20(4,5)23-21/h11-14H,6-10H2,1-5H3

InChI key

XTTRNSNHDCYSEL-UHFFFAOYSA-N

Application

Reagent use for
  • Suzuki-Miyaura cross-coupling reactions and shape-shifting in contorted dibenzotetrathienocoronenes
  • Oligothiophene self-assembly induction into fibers with tunable shape and function
  • Stille coupling and p-conjugated packing structure and hole mobility of bithiophene-bithiazole copolymers with alkyl-thiophene side chains

Reagent used in Preparation of
  • Solution-processed ambipolar field-effect transistor
  • Light harvesting small molecules for use in solution-processed small molecule bulk heterojunction solar cell devices
  • Light-emitting diode (OLED) materials
  • Unsymmetric substituted benzothiadiazole-containing vinyl monomers for RAFT polymerization
  • Pd-catalyzed condensations and synthesis of isoindigo-based oligothiophenes for molecuar bulk heterojunction solar cells
  • Thiophene-benzothiadiazole based donor-acceptor-donor materials

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benzothiadiazole-Containing Pendant Polymers Prepared by RAFT and Their Electro-Optical Properties
Haussler, M.; et al.
Macromolecules, 43, 7101-7110 (2010)
A modular molecular framework for utility in small-molecule solution-processed organic photovoltaic devices
Welch, G. C.; et al.
Journal of Materials Chemistry, 21, 12700-12709 (2011)
Solution-Processed Ambipolar Field-Effect Transistor Based on Diketopyrrolopyrrole Functionalized with Benzothiadiazole
Zhang, Y.; et al.
Advances in Functional Materials, 22, 97-105 (2012)
Francesca Di Maria et al.
Journal of the American Chemical Society, 133(22), 8654-8661 (2011-04-30)
Functional supramolecular architectures for bottom-up organic nano- and microtechnology are a high priority research topic. We discovered a new recognition algorithm, resulting from the combination of thioalkyl substituents and head-to-head regiochemistry of substitution, to induce the spontaneous self-assembly of sulfur
Bithiophene-bithiazole alternating copolymers with thiophene side chains: Synthesis by organometallic polycondensation and chemical properties of the copolymers
Yamamoto, T.; et al.
Journal of Polymer Science Part A: Polymer Chemistry, 49, 1508-1512 (2011)

Articles

Oligothiophenes are important organic electronic materials which can be produced using synthetic intermediates and Suzuki coupling.

Suzuki-Miyaura cross-coupling reaction is extensively used in organic chemistry, polymer science, and pharmaceutical industries.

Suzuki-Miyaura cross-coupling reaction is extensively used in organic chemistry, polymer science, and pharmaceutical industries.

Suzuki-Miyaura cross-coupling reaction is extensively used in organic chemistry, polymer science, and pharmaceutical industries.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service