Skip to Content
Merck
All Photos(1)

Key Documents

61745

Sigma-Aldrich

N-Lauroylsarcosine sodium salt

≥97.0% (HPLC)

Synonym(s):

N-Dodecanoyl-N-methylglycine sodium salt, Sarkosyl NL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10CON(CH3)CH2COONa
CAS Number:
Molecular Weight:
293.38
Beilstein:
5322974
EC Number:
MDL number:
UNSPSC Code:
12161902
PubChem Substance ID:
NACRES:
NA.22

description

anionic

Quality Level

Assay

≥97.0% (HPLC)

form

powder

mol wt

micellar avg mol wt 600

aggregation number

2

technique(s)

protein expression: suitable
protein purification: suitable
protein quantification: suitable

impurities

≤7% water

CMC

14.6 mM (20-25°C)

solubility

H2O: very soluble 293 g/L at 20 °C

SMILES string

[Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O

InChI

1S/C15H29NO3.Na/c1-3-4-5-6-7-8-9-10-11-12-14(17)16(2)13-15(18)19;/h3-13H2,1-2H3,(H,18,19);/q;+1/p-1

InChI key

KSAVQLQVUXSOCR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

N-Lauroylsarcosine is an anionic surfactant with an ability to denature proteins. Due to its microbicidal property, N-lauroylsarcosine is being considered as a potent anti-microbicide in topical formulations, especially against sexually transmitted diseases (STDs).

Application

N-Lauroylsarcosine sodium salt has been used to study the complexing ability of a novel amphiphilic γ-cyclodextrin with anionic surfactants.

Other Notes

Anionic surfactant.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

512.6 °F - closed cup

Flash Point(C)

267 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A facile one-pot synthesis of novel amphiphilic perfluoroalkyl ester functionalized ?-cyclodextrin and complex formation with anionic surfactants.
Lim KT
Journal of Fluorine Chemistry, 127(6), 730-735 (2006)
Gültekin Tamgüney et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(35), 15002-15006 (2009-08-27)
Prions are infectious proteins that cause fatal neurodegenerative diseases. Because astrocytic gliosis marked by the deposition of fibrils composed of GFAP is a prominent feature of prion disease, we asked whether GFAP might be used as a surrogate marker for
Linda A Terry et al.
Veterinary research, 42, 65-65 (2011-05-20)
Classical scrapie is a naturally transmitted prion disease of sheep and goats. Contaminated environments may contribute to the spread of disease and evidence from animal models has implicated urine, blood, saliva, placenta and faeces as possible sources of the infection.
Jocelyne Piret et al.
Antimicrobial agents and chemotherapy, 46(9), 2933-2942 (2002-08-17)
The mechanisms of herpes simplex virus (HSV) inactivation by sodium lauryl sulfate (SLS) and n-lauroylsarcosine (LS), two anionic surfactants with protein denaturant potency, have been evaluated in cultured cells. Results showed that pretreatment of HSV type 1 (HSV-1) strain F
S Roy et al.
Antimicrobial agents and chemotherapy, 45(6), 1671-1681 (2001-05-17)
The microbicidal efficacies of two anionic surfactants, sodium lauryl sulfate (SLS) and n-lauroylsarcosine (LS), were evaluated in cultured cells and in a murine model of herpes simplex type 2 (HSV-2) intravaginal infection. In vitro studies showed that SLS and LS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service