Skip to Content
Merck
All Photos(1)

Key Documents

61250

Sigma-Aldrich

DL-Kynurenine

≥95.0% (NT), for peptide synthesis

Synonym(s):

2-Amino-3-(2-aminobenzoyl)propionic acid, 2-Amino-4-(2-aminophenyl)-4-oxobutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2O3
CAS Number:
Molecular Weight:
208.21
Beilstein:
2697333
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Product Name

DL-Kynurenine, ≥95.0% (NT)

Quality Level

Assay

≥95.0% (NT)

form

powder

mp

~235 °C (dec.)

application(s)

peptide synthesis

SMILES string

NC(CC(=O)c1ccccc1N)C(O)=O

InChI

1S/C10H12N2O3/c11-7-4-2-1-3-6(7)9(13)5-8(12)10(14)15/h1-4,8H,5,11-12H2,(H,14,15)

InChI key

YGPSJZOEDVAXAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam K Walker et al.
Molecular psychiatry, 24(10), 1523-1532 (2018-07-11)
Inflammation activates indoleamine 2,3-dioxygenase (IDO) which metabolizes tryptophan into kynurenine. Circulating kynurenine is transported into the brain by the large amino transporter LAT1 at the level of the blood-brain barrier. We hypothesized that administration of leucine that has a high
Alban Bessede et al.
Nature, 511(7508), 184-190 (2014-06-17)
Disease tolerance is the ability of the host to reduce the effect of infection on host fitness. Analysis of disease tolerance pathways could provide new approaches for treating infections and other inflammatory diseases. Typically, an initial exposure to bacterial lipopolysaccharide
Rahul Shinde et al.
Nature immunology, 19(6), 571-582 (2018-05-16)
The transcription factor AhR modulates immunity at multiple levels. Here we report that phagocytes exposed to apoptotic cells exhibited rapid activation of AhR, which drove production of the cytokine IL-10. Activation of AhR was dependent on interactions between apoptotic-cell DNA
Adam Kosti et al.
Genome biology, 21(1), 195-195 (2020-08-09)
RNA-binding proteins (RBPs) function as master regulators of gene expression. Alterations in RBP expression and function are often observed in cancer and influence critical pathways implicated in tumor initiation and growth. Identification and characterization of oncogenic RBPs and their regulatory
Dag Holmberg et al.
Malaria journal, 16(1), 303-303 (2017-07-30)
One-fourth of children with cerebral malaria (CM) retain cognitive sequelae up to 2 years after acute disease. The kynurenine pathway of the brain, forming neuroactive metabolites, e.g. the NMDA-receptor antagonist kynurenic acid (KYNA), has been implicated in long-term cognitive dysfunction in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service