Skip to Content
Merck
All Photos(3)

Key Documents

595675

Sigma-Aldrich

2-Bromo-5-fluoropyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3BrFN
CAS Number:
Molecular Weight:
175.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

bp

80-83 °C/44 mmHg (lit.)

mp

30-31 °C (lit.)

SMILES string

Fc1ccc(Br)nc1

InChI

1S/C5H3BrFN/c6-5-2-1-4(7)3-8-5/h1-3H

InChI key

UODINHBLNPPDPD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Bromo-5-fluoropyridine can be used in the synthesis of the following:
  • 5-fluoro-2-phenylpyridine via Suzuki coupling reaction with phenylboronic acid
  • 5-fluoro-2-(p-tolyl)pyridine via Suzuki coupling reaction with p-tolylboronic acid
It can undergo palladium-catalyzed homo-coupling reaction to give the corresponding biaryl. It can also be employed in a palladium-catayzed α-arylation of esters leading to 4-pyridylcarboxypiperidines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lee K and Lee HP
Tetrahedron Letters, 49(27), 4302-4305 (2008)
"A fast and oxygen-promoted protocol for the ligand-free Suzuki reaction of 2-halogenated pyridines in aqueous media"
Liu C and Yang W
Chemical Communications (Cambridge, England), 41, 6267-6269 (2009)
"A General and Highly Efficient Method for the Construction of Aryl-Substituted N-Heteroarenes"
Liu C, et al.
European Journal of Organic Chemistry, 1010(29), 5548-5551 (2010)
Tetrahedron Letters, 48, 1191-1191 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service