Skip to Content
Merck
All Photos(1)

Key Documents

59470

Sigma-Aldrich

5,6-O-Isopropylidene-L-gulonic acid γ-lactone

≥99.0% (sum of enantiomers, TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14O6
CAS Number:
Molecular Weight:
218.20
Beilstein:
5267697
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (sum of enantiomers, TLC)

optical activity

[α]20/D +62±2°, c = 1% in H2O

mp

166-170 °C

SMILES string

CC1(C)OC[C@H](O1)[C@H]2OC(=O)[C@@H](O)[C@H]2O

InChI

1S/C9H14O6/c1-9(2)13-3-4(15-9)7-5(10)6(11)8(12)14-7/h4-7,10-11H,3H2,1-2H3/t4-,5+,6-,7+/m0/s1

InChI key

JNTPPVKRHGNFKM-BNHYGAARSA-N

Other Notes

Starting material for the synthesis of an array of compounds modified in the positions 2 and 3; synthesis of isopropylidene-L-glyceraldehyde

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C. Hubschwerlen et al.
Organic Syntheses, 72, 1-1 (1995)
The Journal of Organic Chemistry, 52, 1093-1093 (1987)
J.A. Vekemans et al.
Rec. Trav. Chim., 104, 266-266 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service