Skip to Content
Merck
All Photos(1)

Key Documents

594032

Sigma-Aldrich

Xenon-132Xe

99.5 atom %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
132Xe
CAS Number:
Molecular Weight:
131.90
MDL number:
UNSPSC Code:
12142001
PubChem Substance ID:

isotopic purity

99.5 atom %

Assay

≥99.5% (Total Xenon)

form

gas

impurities

<0.5% Total others

bp

-107.1 °C (lit.)

mass shift

depleted

SMILES string

[132Xe]

InChI

1S/Xe/i1+1

InChI key

FHNFHKCVQCLJFQ-OUBTZVSYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mathias Steglich et al.
The Journal of chemical physics, 142(24), 244311-244311 (2015-07-03)
The 1(3)Σu (-)←X(3)Σg (-) transition of linear HC5H (A) has been observed in a neon matrix and gas phase. The assignment is based on mass-selective experiments, extrapolation of previous results of the longer HC2n+1H homologues, and density functional and multi-state
Mu He et al.
Magnetic resonance imaging, 33(7), 877-885 (2015-05-06)
The aim of this study was to evaluate the effect of hyperpolarized (129)Xe dose on image signal-to-noise ratio (SNR) and ventilation defect conspicuity on both multi-slice gradient echo and isotropic 3D-radially acquired ventilation MRI. Ten non-smoking older subjects (ages 60.8±7.9years)
Arghya Chakraborty et al.
The Journal of chemical physics, 143(8), 084312-084312 (2015-09-04)
The electronic transitions of 9-fluorenone FL(+) and 2,3,6,7-dibenzotropone DBT(+) cations were detected in 6 K neon matrices following a mass-selective deposition. The absorptions at 649.2 and 472.2 nm are assigned to the 2 (2)B1←X̃(2)A2 FL(+) and 2(2)A(')←X̃(2)A(') DBT(+) transitions. Absorption spectra of
Celia van der Merwe et al.
Biochemical and biophysical research communications, 447(2), 334-340 (2014-04-12)
Parkinson's disease (PD), defined as a neurodegenerative disorder, is characterized by the loss of dopaminergic neurons in the substantia nigra in the midbrain. Loss-of-function mutations in the parkin gene are a major cause of autosomal recessive, early-onset PD. Parkin has
Eladio Rodriguez-Diaz et al.
Inflammatory bowel diseases, 20(6), 1029-1036 (2014-05-07)
In 10% to 15% of individuals, inflammatory bowel disease (IBD) is difficult to classify as ulcerative colitis (UC) or Crohn's disease (CD). Previous work has demonstrated that probe-based elastic scattering spectroscopy (ESS) can produce spectra, informed by parameters like tissue

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service