Skip to Content
Merck
All Photos(1)

Key Documents

589292

Sigma-Aldrich

Lithocholic acid-11,12-d2

97 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24D2H38O3
CAS Number:
Molecular Weight:
378.58
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

97 atom % D

Quality Level

mp

184-186 °C (lit.)

mass shift

M+2

SMILES string

[H][C@]12CC[C@@]3([H])[C@]4([H])CC[C@H]([C@H](C)CCC(O)=O)[C@@]4(C)C([2H])C([2H])[C@]3([H])[C@@]1(C)CC[C@@H](O)C2

InChI

1S/C24H40O3/c1-15(4-9-22(26)27)19-7-8-20-18-6-5-16-14-17(25)10-12-23(16,2)21(18)11-13-24(19,20)3/h15-21,25H,4-14H2,1-3H3,(H,26,27)/t15-,16-,17-,18+,19-,20+,21+,23+,24-/m1/s1/i11D,13D/t11?,13?,15-,16-,17-,18+,19-,20+,21+,23+,24-

InChI key

SMEROWZSTRWXGI-YBRLEOEUSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service