Skip to Content
Merck
All Photos(1)

Key Documents

579122

Sigma-Aldrich

4-(Trifluoromethyl)cyclohexanecarboxylic acid, mixture of cis and trans

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F3CC6H10CO2H
CAS Number:
Molecular Weight:
196.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

71-75 °C (lit.)

SMILES string

OC(=O)C1CCC(CC1)C(F)(F)F

InChI

1S/C8H11F3O2/c9-8(10,11)6-3-1-5(2-4-6)7(12)13/h5-6H,1-4H2,(H,12,13)

InChI key

LMEAZIIFLVDISW-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service