Skip to Content
Merck
All Photos(2)

Documents

572667

Sigma-Aldrich

Baicalin

95%

Synonym(s):

5,6-Dihydroxy-4-oxo-2-phenyl-4H-1-benzopyran-7-yl-β-D-glucopyranosiduronic acid, Baicalein 7-O-β-D-glucuronide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18O11
CAS Number:
Molecular Weight:
446.36
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

optical activity

[α]20/D −85°, c = 1 in DMSO

mp

202-205 °C (dec.) (lit.)

SMILES string

O[C@@H]1[C@@H](O)[C@@H](O[C@@H]([C@H]1O)C(O)=O)Oc2cc3OC(=CC(=O)c3c(O)c2O)c4ccccc4

InChI

1S/C21H18O11/c22-9-6-10(8-4-2-1-3-5-8)30-11-7-12(14(23)15(24)13(9)11)31-21-18(27)16(25)17(26)19(32-21)20(28)29/h1-7,16-19,21,23-27H,(H,28,29)/t16-,17-,18+,19-,21+/m0/s1

InChI key

IKIIZLYTISPENI-ZFORQUDYSA-N

Gene Information

mouse ... Hexa(15211)

Looking for similar products? Visit Product Comparison Guide

General description

Baicalin is a key flavonoid found in the roots of Scutellaria baicalensis. It is known to have different biological activities, such as anti-oxidative, anti-inflammatory, antitumor and anti-apoptotic activities.

Application

Baicalin has been used in a study to examine its neuroprotective effect in chronically stressed rats.2

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baicalin Attenuates Hypoxia-Induced Pulmonary Arterial Hypertension to Improve Hypoxic Cor Pulmonale by Reducing the Activity of the p38 MAPK Signaling Pathway and MMP-9.
Yan S, et al.
Evidence-Based Complementary and Alternative Medicine : ECAM, 2016 (2016)
Baicalin influences the dendritic morphology of newborn neurons in the hippocampus of chronically stressed rats.
Jiang X, et al.
Neural Regeneration Research, 8(6), 496-496 (2013)
Peng-Fei Yue et al.
International journal of pharmaceutics, 443(1-2), 279-287 (2013-01-08)
Baicalin nanosuspensions, stabilized with 10% TPGS (relative to the weight of baicalin), were transformed into nanosuspensions powders by solidification process. Solidification methods for this transformation included freeze-drying, spray drying or vacuum drying. High pressure homogenization was applied for production of
Peng-Wei Zhuang et al.
CNS neuroscience & therapeutics, 19(3), 154-162 (2013-01-11)
Recent studies revealed that baicalin, a flavonoid compound derived from the root of Scutellaria baicalensis Georgi, could promote neuron differentiation of NSPCs after commencing the differentiation process in vitro. However, this may not be the most efficacious strategy to determinate
Hong-Zhi Wang et al.
The Journal of pharmacology and experimental therapeutics, 350(2), 435-443 (2014-06-05)
This study focused on the potential therapeutic effect of baicalin on collagen-induced arthritis (CIA) in rats and the underlying mechanisms. The CIA rats were injected with baicalin (50, 100, or 200 mg/kg) once daily for 30 days. The rats were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service