Skip to Content
Merck
All Photos(1)

Key Documents

56951

Sigma-Aldrich

N-(2-Aminoethyl)maleimide trifluoroacetate salt

≥98% (T), for peptide synthesis

Synonym(s):

2-Maleimidoethylamine trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2O2 · C2HF3O2
CAS Number:
Molecular Weight:
254.16
MDL number:
UNSPSC Code:
12161502
PubChem Substance ID:
NACRES:
NA.22

product name

N-(2-Aminoethyl)maleimide trifluoroacetate salt, ≥95% (HPLC), ≥98% (T)

Assay

≥95% (HPLC)
≥98% (T)

form

powder

reaction suitability

reagent type: cross-linking reagent
reagent type: linker

functional group

amine
maleimide

polymer architecture

functionality: heterobifunctional

SMILES string

OC(C(F)(F)F)=O.O=C1N(CCN)C(C=C1)=O

InChI

1S/C6H8N2O2.C2HF3O2/c7-3-4-8-5(9)1-2-6(8)10;3-2(4,5)1(6)7/h1-2H,3-4,7H2;(H,6,7)

InChI key

YNHKVOGCDPODMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-(2-Aminoethyl)maleimide trifluoroacetate salt is a thiol reactive cross-linking agent used in the preparation of heparin hydrogels

Application

N-(2-Aminoethyl)maleimide trifluoroacetate salt commonly utilized:
  • As a crosslinking agent to form covalent bonds between polymeric precursors
  • In the synthesis of maleimide-functionalized heparin hydrogels by introducing maleimide functional groups to molecules, enabling subsequent bioconjugation
  • In the synthesis of N-(2-{3-Iodobenzoyl}aminoethyl)maleimide and N-(2-{3-(tri-n-Butylstannyl)benzoyl}aminoethyl)maleimide
  • As a building block in the synthesis of maleimide-functionalized cymantrenyl derivatives

Other Notes

Heterobifunctional linker for introducing a maleimide function into the widely used NOSu-modified fluorescent labels.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High resolution bioprinting of multi-component hydrogels
R Zimmermann, et al.
Biofabrication, 11, 045008-045008 (2019)
Synthesis and investigation of a radioiodinated F3 peptide analog as a SPECT tumor imaging radioligand
Bhojani M S, et al.
PLoS ONE, 6(7), e22418-e22418 (2011)
Synthesis and investigation of a radioiodinated F3 peptide analog as a SPECT tumor imaging radioligand
MS Bhojani, et al.
PLoS ONE, 6, e22418-e22418 (2011)
Eduardo Pérez Del Río et al.
Biomaterials, 259, 120313-120313 (2020-08-24)
Recent achievements in the field of immunotherapy, such as the development of engineered T cells used in adoptive cell therapy, are introducing more efficient strategies to combat cancer. Nevertheless, there are still many limitations. For example, these T cells are
Chemoselective peptide functionalization of starPEG-GAG hydrogels
MV Tsurkan, et al.
Bioconjugate Chemistry, 25, 1942-1950 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service