Skip to Content
Merck
All Photos(1)

Documents

555568

Sigma-Aldrich

Ethyl 2,3-dichloropropionate

97%

Synonym(s):

2,3-Dichloropropionic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OCOCH(Cl)CH2Cl
CAS Number:
Molecular Weight:
171.02
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.4480 (lit.)

bp

164-165 °C (lit.)

density

1.241 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(Cl)CCl

InChI

1S/C5H8Cl2O2/c1-2-9-5(8)4(7)3-6/h4H,2-3H2,1H3

InChI key

RNZPQAZETZXKCQ-UHFFFAOYSA-N

General description

Ethyl 2,3-dichloropropionate can be prepared from ethyl acrylate via chlorination.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polarographic Behavior of Unsymmetrical Polyhalogenated Compounds. Polychlorinated Propionitrile and Derivatives1.
Jura WH and Gaul RJ.
Journal of the American Chemical Society, 80(20), 5402-5540 (1958)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service