Skip to Content
Merck
All Photos(2)

Key Documents

55452

Sigma-Aldrich

D-α-Hydroxyisovaleric acid

≥98.0% (T)

Synonym(s):

(R)-2-Hydroxy-3-methylbutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O3
CAS Number:
Molecular Weight:
118.13
Beilstein:
1721139
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥98.0% (T)

form

solid

optical activity

[α]20/D −17±1°, c = 1% in chloroform

mp

64-67 °C

functional group

carboxylic acid
hydroxyl

SMILES string

CC(C)[C@@H](O)C(O)=O

InChI

1S/C5H10O3/c1-3(2)4(6)5(7)8/h3-4,6H,1-2H3,(H,7,8)/t4-/m1/s1

InChI key

NGEWQZIDQIYUNV-SCSAIBSYSA-N

Application

D-α-Hydroxyisovaleric acid may be used in the preparation of biodegradable, optically active and isotactic poly(D-2-hydroxy-3-methylbutanoic acid).[1]

Other Notes

This chiral α-hydroxy acid is used for the synthesis of peptides and depsipeptides.[2] It is further used as chiral building block.[3] Synthesis of α-substituted α-hydroxy acids via dioxolanone intermediates.[4][5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H.-O. Kim et al.
The Journal of Organic Chemistry, 52, 4531-4531 (1987)
R L Johnson
Journal of medicinal chemistry, 23(6), 666-669 (1980-06-01)
The following N-(alpha-hydroxylakanoyl) derivatives of Leu-Val-Phe-OCH3 were synthesized and tested for their ability to inhibit human amniotic renin: D- and L-alpha-hydroxyisocaproyl-Leu-Val-Phe-OCH3, D- and L-alpha-hydroxyisovaleryl-Leu-Val-Phe-OCH3, L-2-hydroxy-3-phenylpropanoyl-Leu-Val-Phe-OCH3, and D- and L-alpha-hydroxyphenylacetyl-Leu-Val-Phe-OCH3. Analysis of the compounds through the use of Dixion plots showed
W. Hartwig et al.
Liebigs Ann. Chem., 1952-1952 (1982)
Hetero-stereocomplex formation between substituted poly (lactic acid) s with linear and branched side chains, poly (l-2-hydroxybutanoic acid) and poly (d-2-hydroxy-3-methylbutanoic acid).
Tsuji H and Hayakawa T.
Polymer, 55(3), 721-726 (2014)
Identification of 3-hydroxyisovalerylcarnitine in the urine of a patient with multiple carboxylase deficiency.
T Nakanishi et al.
Annals of clinical biochemistry, 30 ( Pt 3), 318-320 (1993-05-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service