Skip to Content
Merck
All Photos(2)

Key Documents

544272

Sigma-Aldrich

5-Chlorothiophene-2-sulfonyl chloride

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H2Cl2O2S2
CAS Number:
Molecular Weight:
217.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

refractive index

n20/D 1.586 (lit.)

bp

112-117 °C (lit.)

density

1.623 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

Clc1ccc(s1)S(Cl)(=O)=O

InChI

1S/C4H2Cl2O2S2/c5-3-1-2-4(9-3)10(6,7)8/h1-2H

InChI key

SORSTNOXGOXWAO-UHFFFAOYSA-N

General description

5-Chlorothiophene-2-sulfonyl chloride can be prepared from 2-chlorothiophene by reacting with chlorosulfonic acid in the presence of phosphorus pentachloride.

Application

5-Chlorothiophene-2-sulfonyl chloride may be used in the preparation of:
  • (S)-N-(5-chlorothiophene-2-sulfonyl)-β,β-diethylalaninol
  • N-(3-(1H-pyrazol-1-yl)pyridin-2-yl)-5-chlorothiophene-2-sulfonamide
  • 5-chloro-4-nitrothiophene-2-sulfonyl chloride

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Afjal H Miah et al.
Organic & biomolecular chemistry, 12(11), 1779-1792 (2014-02-12)
A knowledge-based library of aryl 2,3-dichlorophenylsulfonamides was synthesised and screened as human CCR4 antagonists, in order to identify a suitable hit for the start of a lead-optimisation programme. X-ray diffraction studies were used to identify the pyrazole ring as a
C A Hunt et al.
Journal of medicinal chemistry, 37(2), 240-247 (1994-01-21)
3-Aminoalkyl derivatives of thieno[2,3-b][1,4]thiazine-6-sulfonamide were prepared for evaluation as topically active ocular hypotensive agents. The compounds described were found to be excellent in vitro inhibitors of carbonic anhydrase II and in vivo to lower intraocular pressure in three rabbit models
A facile stereoselective total synthesis of (S)-N-(5-chlorothiophene-2-sulfonyl)-?,?-diethylalaninol.
Reddy BN and Sing RP.
Der Pharma Chemica, 7(1), 201-205 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service