Skip to Content
Merck
All Photos(1)

Documents

543136

Sigma-Aldrich

Propargyl acrylate

98%

Synonym(s):

Acrylic acid propargyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCO2CH2C≡CH
CAS Number:
Molecular Weight:
110.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

contains

200 ppm BHT as inhibitor

refractive index

n20/D 1.447 (lit.)

bp

142-143 °C (lit.)

density

0.997 g/mL at 25 °C (lit.)

SMILES string

C=CC(=O)OCC#C

InChI

1S/C6H6O2/c1-3-5-8-6(7)4-2/h1,4H,2,5H2

InChI key

WPBNLDNIZUGLJL-UHFFFAOYSA-N

General description

Propargyl acrylate is a propargyl ester of acrylic acid. It participates in the preparation of lipid nanoparticles for use in siRNA delivery.

Application

Propargyl acrylate may be used to synthesize:
  • linear poly(N-isopropylacrylamide)(PNIPAM)
  • 1-[3-(2-methyl-2-dodecylsulfanylthiocarbonylsulfanylpropionyloxy)propyl]-1H-[1,2,3]triazol-4-ylmethyl acrylate, an acryloyl trithiocarbonate chain transfer agent
  • propranolol-imprinted core–shell nanoparticles (mipCS)

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

109.9 °F - closed cup

Flash Point(C)

43.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyperbranched polymers via RAFT copolymerization of an acryloyl trithiocarbonate
Vogt AP, et al.
Australian Journal of Chemistry, 60.6, 396-399 (2007)
Andreas Hoffmann et al.
Journal of tissue engineering, 8, 2041731417744485-2041731417744485 (2018-01-11)
Stereolithography is one of the most promising technologies for the production of tailored implants. Within this study, we show the results of a new resin formulation for three-dimensional printing which is also useful for subsequent surface functionalization. The class of
Molecularly imprinted magnetic materials prepared from modular and clickable nanoparticles
Xu C, et al.
Journal of Materials Chemistry, 22.15, 7427-7433 (2012)
Multiparametric approach for the evaluation of lipid nanoparticles for siRNA delivery.
Alabi CA, et al.
Proceedings of the National Academy of Sciences of the USA, 110(32), 12881-12886 (2013)
Temperature-induced phase transition of well-defined cyclic poly (N-isopropylacrylamide) s in aqueous solution
Qiu X-P, et al.
Macromolecules, 40.20, 7069- 7071 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service