Skip to Content
Merck
All Photos(1)

Key Documents

510734

Sigma-Aldrich

6-Chloro-3-pyridinecarbonitrile

97%

Synonym(s):

2-Chloro-5-cyanopyridine, 2-Chloropyridine-5-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3ClN2
CAS Number:
Molecular Weight:
138.55
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

116-120 °C (lit.)

functional group

chloro
nitrile

SMILES string

Clc1ccc(cn1)C#N

InChI

1S/C6H3ClN2/c7-6-2-1-5(3-8)4-9-6/h1-2,4H

InChI key

ORIQLMBUPMABDV-UHFFFAOYSA-N

General description

6-Chloro-3-pyridinecarbonitrile is a substituted pyridine.

Application

6-Chloro-3-pyridinecarbonitrile (2-Chloro-5-cyanopyridine) may be used in the preparation of:
  • (6-chloro-3-pyridyl)methylamine
  • 2-(N-methyl-N-isopropylamino)-5-cyanopyridine
  • S-(5-cyano-2-pyridyl)thiouronium chloride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Practical Synthesis of (6-Chloro-3-pyridyl) methylamine by Highly Selective Hydrogenation of 6-Chloro-3-pyridinecarbonitrile with Improved Raney Nickel Catalyst.
Tanaka K, et al.
Bulletin of the Chemical Society of Japan, 73(5), 1227-1231 (2000)
Bachir Latli et al.
Journal of labelled compounds & radiopharmaceuticals (2018-05-17)
1-(4-Fluorophenyl)-1H-pyrazolo[3,4-c]pyridine-4-carboxylic acid (2-methanesulfonyl-pyridin-4-ylmethyl)-amide (1) and its analogs (2) and (3) are potent CCR1 antagonists intended for the treatment of rheumatoid arthritis. The detailed syntheses of these 3 compounds labeled with carbon-13 as well as the preparation of (1) and (2)
Chemotherapeutic agents of the sulphone type. Part V. 2: 5-Disubstituted derivatives of pyridine.
Forrest HS and Walker J.
Journal of the Chemical Society, 1939-1945 (1948)
An experimental test of CN bond twisting in the TICT state: syn-anti photoisomerization in 2-(N-Methyl-N-isopropylamino)-5-cyanopyridine.
Dobkowski J, et al.
Journal of the American Chemical Society, 124(11), 2406-2407 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service