Skip to Content
Merck
All Photos(2)

Documents

510661

Sigma-Aldrich

Yttrium(III) butoxide solution

0.5 M in toluene, ≥99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Y(OC4H9)3
CAS Number:
Molecular Weight:
308.25
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.9% trace metals basis

form

liquid

reaction suitability

core: yttrium
reagent type: catalyst

concentration

0.5 M in toluene

refractive index

n20/D 1.495

bp

109 °C

density

0.9 g/mL at 25 °C

SMILES string

CCCCO[Y](OCCCC)OCCCC

InChI

1S/3C4H9O.Y/c3*1-2-3-4-5;/h3*2-4H2,1H3;/q3*-1;+3

InChI key

PCOPFSXTYFFNIG-UHFFFAOYSA-N

Application

Precursor to composite film electrode for solid oxide fuel cells.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

46.4 °F - closed cup

Flash Point(C)

8 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service