Skip to Content
Merck
All Photos(3)

Key Documents

498270

Sigma-Aldrich

1-Chloro-N,N,2-trimethyl-1-propenylamine

96%

Synonym(s):

Ghosez′s reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=C(Cl)N(CH3)2
CAS Number:
Molecular Weight:
133.62
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.453 (lit.)

bp

129-130 °C (lit.)

density

1.01 g/mL at 25 °C (lit.)

functional group

amine
chloro

storage temp.

2-8°C

SMILES string

CN(C)\C(Cl)=C(\C)C

InChI

1S/C6H12ClN/c1-5(2)6(7)8(3)4/h1-4H3

InChI key

GQIRIWDEZSKOCN-UHFFFAOYSA-N

Application

1-Chloro-N,N,2-trimethyl-1-propenylamine is an acid halogenation reagent developed by Ghosez, that enables the conversion of carboxylic acids into the corresponding chlorides under strictly neutral conditions. This method was successfully used in the total synthesis of caloporoside, roseophilin, (-)-enniatin B, (±)-epimeloscine and (±)-meloscine.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

80.0 °F - closed cup

Flash Point(C)

26.67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. Bohme and H. G. Viehe, Eds.
Adv. Org. Chem., 9, 421-421 (1976)
A short total synthesis of (?)-epimeloscine and (?)-meloscine enabled by a cascade radical annulation of a divinylcyclopropane.
Zhang H, et al.
Journal of the American Chemical Society, 133(27), 10376-10378 (2011)
Total synthesis of caloporoside.
Furstner A, et al.
The Journal of Organic Chemistry, 63, 3072-3072 (1998)
?-Chloro enamines, reactive intermediates for synthesis: 1-Chloro-N,N,2-trimethylpropenylamine.
Haveaux B, et al.
Organic Syntheses, 59, 26-26 (1980)
A total synthesis of the ammonium ionophore,(?)-enniatin B.
Hu D, et al.
Tetrahedron Letters, 53(32), 4077-4079 (2012)

Articles

Fluoroamidinium salts advance acid halogenation with greater stability and no oxazolones conversion compared to chlorides.

Fluoroamidinium salts advance acid halogenation with greater stability and no oxazolones conversion compared to chlorides.

Fluoroamidinium salts advance acid halogenation with greater stability and no oxazolones conversion compared to chlorides.

Fluoroamidinium salts advance acid halogenation with greater stability and no oxazolones conversion compared to chlorides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service