Skip to Content
Merck
All Photos(1)

Key Documents

494615

Sigma-Aldrich

Piperidine, polymer-bound

extent of labeling: 3.0-4.0 mmol/g loading, 1 % cross-linked with divinylbenzene

Synonym(s):

(Piperidinomethyl)polystyrene, 1-Benzylpiperidine polymer-bound, Copolymer of styrene and divinylbenzene, piperidinomethylated, Polystyrene crosslinked with divinylbenzene, piperidinomethylated

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22

crosslinking

1 % cross-linked with divinylbenzene

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: proton reactive

extent of labeling

3.0-4.0 mmol/g loading

Looking for similar products? Visit Product Comparison Guide

Application

This polymer-bound tertiary amine eases the purification step of organic synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R.J. Booth et al.
Journal of the American Chemical Society, 119, 4882-4882 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service